missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proline, USP, 98.5-101.5%, Spectrum™ Chemical
P1434, 147-85-3, C5H9NO2
$182.33 - $13,909.48
Chemical Identifiers
| CAS | 147-85-3 |
|---|---|
| Molecular Formula | C5H9NO2 |
| Molecular Weight (g/mol) | 115.13 |
| MDL Number | MFCD00064318 |
| InChI Key | ONIBWKKTOPOVIA-UHFFFAOYNA-N |
| IUPAC Name | pyrrolidine-2-carboxylic acid |
| SMILES | OC(=O)C1CCCN1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
18605862
|
N/A | 25 g | Poly Bottle |
Each for $182.33
|
|
||||
|
18605863
|
N/A | 100 g | Poly Bottle |
Each for $337.23
|
|
||||
|
18605864
|
N/A | 1 kg | Poly Bottle |
Each for $1,890.31
|
|
||||
|
18605865
|
N/A | 5 kg | Poly Pail |
Each for $7,398.65
|
|
||||
|
18605866
|
N/A | 25 kg | Fiber Drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical Proline, USP - All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
| 147-85-3 | |
| 115.13 | |
| ONIBWKKTOPOVIA-UHFFFAOYNA-N | |
| OC(=O)C1CCCN1 |
| C5H9NO2 | |
| MFCD00064318 | |
| pyrrolidine-2-carboxylic acid |
Specifications
| 147-85-3 | |
| 0.004 | |
| 0.004 | |
| MFCD00064318 | |
| 0.0003 | |
| OC(=O)C1CCCN1 | |
| 115.13 | |
| USP |
| 1 | |
| 30 ppm | |
| C5H9NO2 | |
| -84.3° to -86.3° | |
| ONIBWKKTOPOVIA-UHFFFAOYNA-N | |
| pyrrolidine-2-carboxylic acid | |
| 98.5101.5% |