missing translation for 'onlineSavingsMsg'
Learn More

Proline, USP, 98.5-101.5%, Spectrum™ Chemical

Catalog No. 18605862
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
5 kg
25 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18605862 25 g Poly Bottle
18605863 100 g Poly Bottle
18605864 1 kg Poly Bottle
18605865 5 kg Poly Pail
18605866 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18605862 Supplier Spectrum Chemical Mfg Cor Supplier No. P143425GM
Get Up to $500 in Chemicals with Purchase
View all promotions

P1434, 147-85-3, C5H9NO2

Spectrum™ Chemical Proline, USP - All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 147-85-3
Molecular Formula C5H9NO2
Molecular Weight (g/mol) 115.13
MDL Number MFCD00064318
InChI Key ONIBWKKTOPOVIA-UHFFFAOYNA-N
IUPAC Name pyrrolidine-2-carboxylic acid
SMILES OC(=O)C1CCCN1

Specifications

CAS Max % 1
Ignition Residue 0.004
Iron (Fe) 30 ppm
Loss on Drying 0.004
Packaging Poly Bottle
Specific Rotation -84.3° to -86.3°
Sulfate 0.0003
Quantity 25 g
Percent Purity 98.5101.5%
Grade USP