Learn More
Sartorius Proline Plus Mechanical Pipette Multipacks
Multipacks combine a set of Proline Plus single-channel mechanical pipettes with matching tip trays and useful accessories. Sartorius Proline Pus Mechanical Pipette Multipacks provides best alternative to replace your used mechanical pipettes.
Supplier: Sartorius LH728672
Includes: 4 x Single-channel pipets (0.5 to 10μL, 2 to 20μL, 20 to 200μL, 100 to 1000μL), Tip Racks of 96 tips (0.1 to 10μL, 0.5 to 200μL, 10 to 1000μL), 1 x Linear Stand and 4 x Pipet Holders
Description
- Low pipetting forces
- Ergonomic finger hook
- Easy volume setting with click stop mechanism
- Quick and easy-to-read volume display minimises risk of volume setting errors
- Thermal insulation of internal components improves accuracy
- Easy to use 'do-it-yourself' calibration
- Easy to clean with only three parts to disassemble
- Fully autoclavable
- High chemical and UV-resistance
- High-quality standard Optifit tips for various applications
- Optiload tip loading mechanism allows tip loading with an equal constant force onto every channel and enables perfect tip sealing onto every individual tip cone
Warranty
Two years
Chemical Identifiers
| Optifit Tips | |
| Reusable | |
| Proline Plus | |
| Mechanical Pipette Multipacks | |
| UV resistant |
| Autoclavable | |
| Proline Plus | |
| Non-sterile | |
| Safe-Cone Filter | |
| 2 Years |
Specifications
| Optifit Tips | |
| Reusable | |
| 1 | |
| Non-sterile | |
| 0.5 to 10 μL, 2 to 20 μL, 20 to 200 μL, 100 to 1000 μL | |
| 4 x Single-channel pipets (0.5 to 10μL, 2 to 20μL, 20 to 200μL, 100 to 1000μL), Tip Racks of 96 tips (0.1 to 10μL, 0.5 to 200μL, 10 to 1000μL), 1 x Linear Stand and 4 x Pipet Holders | |
| UV resistant |
| Autoclavable | |
| Proline Plus | |
| Proline Plus | |
| Mechanical Pipette Multipacks | |
| Safe-Cone Filter | |
| 10/Pk. | |
| 2 Years |