missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Probucol | |
| 128°C | |
| C31H48O2S2 | |
| 25 g | |
| FYPMFJGVHOHGLL-UHFFFAOYSA-N | |
| 2,6-ditert-butyl-4-[2-(3,5-ditert-butyl-4-hydroxyphenyl)sulfanylpropan-2-ylsulfanyl]phenol | |
| 4912 | |
| 516.84 | |
| Crystalline Powder |
| 23288-49-5 | |
| White-Yellow | |
| MFCD00079281 | |
| probucol, lorelco, biphenabid, bisphenabid, lurselle, bisbid, lesterol, lursell, panavir, probucolum | |
| CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)SC(C)(C)SC2=CC(=C(C(=C2)C(C)(C)C)O)C(C)(C)C | |
| 516.843 | |
| CHEBI:8427 | |
| ≥98.0% (HPLC) |
Chemical Identifiers
| 23288-49-5 | |
| 516.843 | |
| FYPMFJGVHOHGLL-UHFFFAOYSA-N | |
| 4912 | |
| 2,6-ditert-butyl-4-[2-(3,5-ditert-butyl-4-hydroxyphenyl)sulfanylpropan-2-ylsulfanyl]phenol |
| C31H48O2S2 | |
| MFCD00079281 | |
| probucol, lorelco, biphenabid, bisphenabid, lurselle, bisbid, lesterol, lursell, panavir, probucolum | |
| CHEBI:8427 | |
| CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)SC(C)(C)SC2=CC(=C(C(=C2)C(C)(C)C)O)C(C)(C)C |
Safety and Handling
RTECSNumber : AL3705000
TSCA : No