missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Probucol 98.0+%, TCI America™
$115.51 - $342.69
Chemical Identifiers
| CAS | 23288-49-5 |
|---|---|
| Molecular Formula | C31H48O2S2 |
| Molecular Weight (g/mol) | 516.843 |
| MDL Number | MFCD00079281 |
| InChI Key | FYPMFJGVHOHGLL-UHFFFAOYSA-N |
| Synonym | probucol, lorelco, biphenabid, bisphenabid, lurselle, bisbid, lesterol, lursell, panavir, probucolum |
| PubChem CID | 4912 |
| ChEBI | CHEBI:8427 |
| IUPAC Name | 2,6-ditert-butyl-4-[2-(3,5-ditert-butyl-4-hydroxyphenyl)sulfanylpropan-2-ylsulfanyl]phenol |
| SMILES | CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)SC(C)(C)SC2=CC(=C(C(=C2)C(C)(C)C)O)C(C)(C)C |
Chemical Identifiers
| 23288-49-5 | |
| 516.843 | |
| FYPMFJGVHOHGLL-UHFFFAOYSA-N | |
| 4912 | |
| 2,6-ditert-butyl-4-[2-(3,5-ditert-butyl-4-hydroxyphenyl)sulfanylpropan-2-ylsulfanyl]phenol |
| C31H48O2S2 | |
| MFCD00079281 | |
| probucol, lorelco, biphenabid, bisphenabid, lurselle, bisbid, lesterol, lursell, panavir, probucolum | |
| CHEBI:8427 | |
| CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)SC(C)(C)SC2=CC(=C(C(=C2)C(C)(C)C)O)C(C)(C)C |
Specifications
| 23288-49-5 | |
| White-Yellow | |
| MFCD00079281 | |
| probucol, lorelco, biphenabid, bisphenabid, lurselle, bisbid, lesterol, lursell, panavir, probucolum | |
| CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)SC(C)(C)SC2=CC(=C(C(=C2)C(C)(C)C)O)C(C)(C)C | |
| 516.843 | |
| CHEBI:8427 | |
| ≥98.0% (HPLC) | |
| Probucol |
| 128°C | |
| C31H48O2S2 | |
| 5 g | |
| FYPMFJGVHOHGLL-UHFFFAOYSA-N | |
| 2,6-ditert-butyl-4-[2-(3,5-ditert-butyl-4-hydroxyphenyl)sulfanylpropan-2-ylsulfanyl]phenol | |
| 4912 | |
| 516.84 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : AL3705000
TSCA : No