missing translation for 'onlineSavingsMsg'
Learn More

Promega Corporation ProAlanase, Mass Spec Grade (0.2 μg/μL)

Catalog No. PRVA2161
Encompass
Change view
Click to view available options
Quantity:
5 μg, 15 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRVA2161 5 μg, 15 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRVA2161 Supplier Promega Corporation Supplier No. VA2161

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The new site-specific endoprotease targets proline and alanine residues, cleaving primarily C-terminal to these residues. Active at acidic pH (1-5.5) with a pH optimum of ~1.5, and offers short digestion times of 1-2 hours.

  • New site-specific endoprotease that targets proline and alanine residues.
  • Cleaves primarily C-terminal to proline and alanine residues.
  • Active at acidic pH (1-5.5) with a pH optimum of ~1.5.
  • Short digestion times of 1-2 hours.
  • High specificity and efficiency.
  • Consistent and reproducible results.
  • Suitable for various proteomics applications.
  • Compatible with existing protocols.
  • Easy integration into existing workflows.
  • Reliable performance for precise protein digestion.

Specifications

For Use With (Equipment) Mass Spectrometer
Solubility Soluble in digestion buffers
Product Type Protease
Starting Material Protein Samples
Final Product Type Digested peptides
Detection Method Fluorescence
Format Bottle
Workflow Step Protein digestion
Quantity 5 μg, 15 μg
For Use With (Application) Protein digestion for mass spectrometry analysis
Form Lyophilized Powder
Show More Show Less