missing translation for 'onlineSavingsMsg'
Learn More

Genlantis™ PrimaPure™ Human Dermal Microvascular Endothelial Cells (HMVEC) and Kits

Functionally characterized

$2,616.52 - $3,729.60

Specifications

Cell Type Adult HDMEC
For Use With (Application) For the study of angiogenesis, tumor metastasis, wound healing, inflammation, macromolecule transport, drug metabolism
Media Recommendation HMVEC Growth Medium
Source Human Neonatal Foreskin or Adult Skin
Sufficient For 16 Population Doublings
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PH10005A
Encompass
View Documents Promotion Details
Genlantis™
PH10005A
>500000 Cells
Each for $2,616.52
Only null left
 
PH10005AK
Encompass
View Documents Promotion Details
Genlantis™
PH10005AK
1 Kit
Each for $3,729.60
Only null left
 
Description

Description

HMVEC are isolated from normal human neonatal foreskin or adult skin capillaries. They are cryopreserved at second passage and can be cultured and propagated at least 16 population doublings. They are responsive to cytokine stimulation in the expression of cell adhesion molecules. HMVEC express CD36 constitutively, while large vessel endothelial cells do not. HMVEC is an organ-specific endothelial cell system for the study of angiogenesis1, tumor metastasis2,3, wound healing4, inflammation5,6, macromolecule transport7, and drug metabolism8. There is growing evidence of differences in physiology and pharmacology between microvascular endothelial cells and macrovascular endothelial cells9,10.

  • Single donors to allow variability comparisons
  • Each lot tested negative for HIV, Hepatitis B and C, mycoplasma, bacteria, yeast, and fungi
  • Performance guaranteed with Genlantis media and reagents

1. Swerlick, R.A. et.al, J. Invest. Dermatol. 97(2):190 (1991).
2. Folkman, J. et al, Nature 288:551 (1980).
3. Lee, K.H. et al, J. Invest. Dermatol. 98(1):79 (1992).
4. Deryugina, E.I. et al, Hybridoma 15(4):279 (1996).
5. Sepp, N.T. et al, J. Invest. Dermatol. 104(2):277 (1995).
6. Swerlick, R.A. et al, J. Invest. Dermatol. 100(1):111S (1993).
7. Hanasaki, K. et al, J. Bio. Chem. 270(13):7533 (1995).
8. Pruckler, J.M. et al, Pathobiology 61:283 (1993).
9. Foster, C.A. et al, J. Dermatol. 21(11):847 (1994).
10. Schnitzer, J.E. et al, BBRC. 199(1):11 (1994).

Specifications

Specifications

Adult HDMEC
HMVEC Growth Medium
16 Population Doublings
For the study of angiogenesis, tumor metastasis, wound healing, inflammation, macromolecule transport, drug metabolism
Human Neonatal Foreskin or Adult Skin
Videos
SDS
Documents

Documents