missing translation for 'onlineSavingsMsg'
Learn More

Genlantis™ PrimaPure™ Human Dermal Microvascular Endothelial Cells (HMVEC) and Kits

Catalog No. PH10005AK
Encompass
Change view
Click to view available options
Quantity:
>500000 Cells
1 Kit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PH10005AK 1 Kit
PH10005A >500000 Cells
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PH10005AK Supplier Genlantis™ Supplier No. PH10005AK

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Functionally characterized

HMVEC are isolated from normal human neonatal foreskin or adult skin capillaries. They are cryopreserved at second passage and can be cultured and propagated at least 16 population doublings. They are responsive to cytokine stimulation in the expression of cell adhesion molecules. HMVEC express CD36 constitutively, while large vessel endothelial cells do not. HMVEC is an organ-specific endothelial cell system for the study of angiogenesis1, tumor metastasis2,3, wound healing4, inflammation5,6, macromolecule transport7, and drug metabolism8. There is growing evidence of differences in physiology and pharmacology between microvascular endothelial cells and macrovascular endothelial cells9,10.

  • Single donors to allow variability comparisons
  • Each lot tested negative for HIV, Hepatitis B and C, mycoplasma, bacteria, yeast, and fungi
  • Performance guaranteed with Genlantis media and reagents

1. Swerlick, R.A. et.al, J. Invest. Dermatol. 97(2):190 (1991).
2. Folkman, J. et al, Nature 288:551 (1980).
3. Lee, K.H. et al, J. Invest. Dermatol. 98(1):79 (1992).
4. Deryugina, E.I. et al, Hybridoma 15(4):279 (1996).
5. Sepp, N.T. et al, J. Invest. Dermatol. 104(2):277 (1995).
6. Swerlick, R.A. et al, J. Invest. Dermatol. 100(1):111S (1993).
7. Hanasaki, K. et al, J. Bio. Chem. 270(13):7533 (1995).
8. Pruckler, J.M. et al, Pathobiology 61:283 (1993).
9. Foster, C.A. et al, J. Dermatol. 21(11):847 (1994).
10. Schnitzer, J.E. et al, BBRC. 199(1):11 (1994).

Specifications

Content And Storage 4° and -20°C
Cell Type Adult HDMEC
For Use With (Application) For the study of angiogenesis, tumor metastasis, wound healing, inflammation, macromolecule transport, drug metabolism
Includes One ampoule of cryopreserved HMVEC, 500mL of HMVEC cell growth medium, one subculture reagent kit (100mL each of HEPES buffered saline solution, trypsin/EDTA, and trypsin neutralizing solution)
Media Recommendation HMVEC Growth Medium
Quantity 1 Kit
Source Human Neonatal Foreskin or Adult Skin
Sufficient For 16 Population Doublings