Learn More
Prilocaine hydrochloride, 98%
A local anesthetic | CAS: 1786-81-8 | C13H21ClN2O | 256.774 g/mol
Supplier: Thermo Scientific Chemicals J6064809
Description
A local anesthetic
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Prilocaine hydrochloride | |
| 167°C to 168°C | |
| C13H21ClN2O | |
| 10 g | |
| prilocaine hydrochloride, propitocaine hydrochloride, prilocaine hcl, xylonest, prilocaine chloride, citanest hydrochloride, citanest plain, n-2-methylphenyl-2-propylamino propanamide hydrochloride, 2-propylamino | |
| CCCNC(C)C(=O)NC1=CC=CC=C1C.Cl | |
| 256.774 | |
| CHEBI:32053 | |
| 98% |
| 1786-81-8 | |
| White | |
| MFCD00079279 | |
| 14,7743 | |
| BJPJNTKRKALCPP-UHFFFAOYSA-N | |
| N-(2-methylphenyl)-2-(propylamino)propanamide;hydrochloride | |
| 92163 | |
| 256.77 | |
| Powder |
Chemical Identifiers
| 1786-81-8 | |
| 256.774 | |
| BJPJNTKRKALCPP-UHFFFAOYSA-N | |
| 92163 | |
| N-(2-methylphenyl)-2-(propylamino)propanamide;hydrochloride |
| C13H21ClN2O | |
| MFCD00079279 | |
| prilocaine hydrochloride, propitocaine hydrochloride, prilocaine hcl, xylonest, prilocaine chloride, citanest hydrochloride, citanest plain, n-2-methylphenyl-2-propylamino propanamide hydrochloride, 2-propylamino | |
| CHEBI:32053 | |
| CCCNC(C)C(=O)NC1=CC=CC=C1C.Cl |
Safety and Handling
EINECSNumber : 217-244-0
RTECSNumber : UG5775000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only