missing translation for 'onlineSavingsMsg'
Learn More

Prilocaine hydrochloride, 98%

Catalog No. p-7041898
Change view
Click to view available options
Quantity:
1 g
10 g
5 g
Catalog No. Quantity
AAJ6064803 1 g
AAJ6064806 5 g
AAJ6064809 10 g
3 options

Catalog No. AAJ6064803

Supplier: Thermo Scientific Chemicals J6064803

Only null left

A local anesthetic | CAS: 1786-81-8 | C13H21ClN2O | 256.774 g/mol

A local anesthetic

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1786-81-8
Molecular Formula C13H21ClN2O
Molecular Weight (g/mol) 256.774
MDL Number MFCD00079279
InChI Key BJPJNTKRKALCPP-UHFFFAOYSA-N
Synonym prilocaine hydrochloride, propitocaine hydrochloride, prilocaine hcl, xylonest, prilocaine chloride, citanest hydrochloride, citanest plain, n-2-methylphenyl-2-propylamino propanamide hydrochloride, 2-propylamino
PubChem CID 92163
ChEBI CHEBI:32053
IUPAC Name N-(2-methylphenyl)-2-(propylamino)propanamide;hydrochloride
SMILES CCCNC(C)C(=O)NC1=CC=CC=C1C.Cl

Specifications

Melting Point 167°C to 168°C
Color White
Quantity 1 g
Merck Index 14,7743
Formula Weight 256.77
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Prilocaine hydrochloride
EINECSNumber 217-244-0
RTECSNumber UG5775000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only