missing translation for 'onlineSavingsMsg'
Learn More

Pretilachlor 97.0+%, TCI America™

Supplier:  TCI America P196725G

 View more versions of this product

Catalog No. P196725G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Pretilachlor
129°C
MFCD00161476
3082
YLPGTOIOYRQOHV-UHFFFAOYSA-N
2-chloro-N-(2,6-diethylphenyl)-N-(2-propoxyethyl)acetamide
91644
311.85
Liquid
51218-49-6
C17H26ClNO2
25 g
pretilachlor, solnet, 2-chloro-n-2,6-diethylphenyl-n-2-propoxyethyl acetamide, rifit, pretilachlore, pretilachlor bsi:iso, pretilachlore iso-french, 2-chloro-2',6'-diethyl-n-2-propoxyethyl acetanilide, unii-617k7lm10d, n-propoxyethyl-n-chloroacetyl-2,6-diethylaniline
CCCOCCN(C1=C(C=CC=C1CC)CC)C(=O)CCl
311.85
CHEBI:34931
≥97.0% (GC)

Chemical Identifiers

51218-49-6
311.85
YLPGTOIOYRQOHV-UHFFFAOYSA-N
91644
2-chloro-N-(2,6-diethylphenyl)-N-(2-propoxyethyl)acetamide
C17H26ClNO2
MFCD00161476
pretilachlor, solnet, 2-chloro-n-2,6-diethylphenyl-n-2-propoxyethyl acetamide, rifit, pretilachlore, pretilachlor bsi:iso, pretilachlore iso-french, 2-chloro-2',6'-diethyl-n-2-propoxyethyl acetanilide, unii-617k7lm10d, n-propoxyethyl-n-chloroacetyl-2,6-diethylaniline
CHEBI:34931
CCCOCCN(C1=C(C=CC=C1CC)CC)C(=O)CCl
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : AB5427000

TSCA : No

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions