CAS RN 51218-49-6
CAS RN 51218-49-6
IUPAC Name:
2-chloro-N-(2,6-diethylphenyl)-N-(2-propoxyethyl)acetamide
Synonyms:
solnet pretilchlor pretilachlore pretilachlor rifit
Molecular Weight (g/mol):
311.85
Molecular Formula:
C17H26ClNO2
InChi Key:
YLPGTOIOYRQOHV-UHFFFAOYSA-N
SMILES:
CCCOCCN(C(=O)CCl)C1=C(CC)C=CC=C1CC
1
–
1
of
1
results
1
Pretilachlor 97.0+%, TCI America™
CAS: 51218-49-6 Molecular Formula: C17H26ClNO2 Molecular Weight (g/mol): 311.85 MDL Number: MFCD00161476 InChI Key: YLPGTOIOYRQOHV-UHFFFAOYSA-N Synonym: pretilachlor,solnet,2-chloro-n-2,6-diethylphenyl-n-2-propoxyethyl acetamide,rifit,pretilachlore,pretilachlor bsi:iso,pretilachlore iso-french,2-chloro-2',6'-diethyl-n-2-propoxyethyl acetanilide,unii-617k7lm10d,n-propoxyethyl-n-chloroacetyl-2,6-diethylaniline PubChem CID: 91644 ChEBI: CHEBI:34931 IUPAC Name: 2-chloro-N-(2,6-diethylphenyl)-N-(2-propoxyethyl)acetamide SMILES: CCCOCCN(C1=C(C=CC=C1CC)CC)C(=O)CCl