missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pretilachlor 97.0+%, TCI America™
$154.71 - $458.56
Chemical Identifiers
| CAS | 51218-49-6 |
|---|---|
| Molecular Formula | C17H26ClNO2 |
| Molecular Weight (g/mol) | 311.85 |
| MDL Number | MFCD00161476 |
| InChI Key | YLPGTOIOYRQOHV-UHFFFAOYSA-N |
| Synonym | pretilachlor, solnet, 2-chloro-n-2,6-diethylphenyl-n-2-propoxyethyl acetamide, rifit, pretilachlore, pretilachlor bsi:iso, pretilachlore iso-french, 2-chloro-2',6'-diethyl-n-2-propoxyethyl acetanilide, unii-617k7lm10d, n-propoxyethyl-n-chloroacetyl-2,6-diethylaniline |
| PubChem CID | 91644 |
| ChEBI | CHEBI:34931 |
| IUPAC Name | 2-chloro-N-(2,6-diethylphenyl)-N-(2-propoxyethyl)acetamide |
| SMILES | CCCOCCN(C1=C(C=CC=C1CC)CC)C(=O)CCl |
Chemical Identifiers
| 51218-49-6 | |
| 311.85 | |
| YLPGTOIOYRQOHV-UHFFFAOYSA-N | |
| 91644 | |
| 2-chloro-N-(2,6-diethylphenyl)-N-(2-propoxyethyl)acetamide |
| C17H26ClNO2 | |
| MFCD00161476 | |
| pretilachlor, solnet, 2-chloro-n-2,6-diethylphenyl-n-2-propoxyethyl acetamide, rifit, pretilachlore, pretilachlor bsi:iso, pretilachlore iso-french, 2-chloro-2',6'-diethyl-n-2-propoxyethyl acetanilide, unii-617k7lm10d, n-propoxyethyl-n-chloroacetyl-2,6-diethylaniline | |
| CHEBI:34931 | |
| CCCOCCN(C1=C(C=CC=C1CC)CC)C(=O)CCl |
Specifications
| 51218-49-6 | |
| C17H26ClNO2 | |
| 5 g | |
| pretilachlor, solnet, 2-chloro-n-2,6-diethylphenyl-n-2-propoxyethyl acetamide, rifit, pretilachlore, pretilachlor bsi:iso, pretilachlore iso-french, 2-chloro-2',6'-diethyl-n-2-propoxyethyl acetanilide, unii-617k7lm10d, n-propoxyethyl-n-chloroacetyl-2,6-diethylaniline | |
| CCCOCCN(C1=C(C=CC=C1CC)CC)C(=O)CCl | |
| 311.85 | |
| CHEBI:34931 | |
| ≥97.0% (GC) | |
| Pretilachlor |
| 129°C | |
| MFCD00161476 | |
| 3082 | |
| YLPGTOIOYRQOHV-UHFFFAOYSA-N | |
| 2-chloro-N-(2,6-diethylphenyl)-N-(2-propoxyethyl)acetamide | |
| 91644 | |
| 311.85 | |
| Liquid |
Safety and Handling
RTECSNumber : AB5427000
TSCA : No
Recommended Storage : Refrigerator