missing translation for 'onlineSavingsMsg'
Learn More

Cytiva PreScission™ Protease

Catalog No. 45001319
Change view
Click to view available options
Quantity:
500U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45001319 500U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45001319 Supplier Cytiva Supplier No. 27084301
Only null left

Use for site-specific separation of the GST tag from proteins expressed using pGEX-6P vectors. Cytiva Life Sciences™ PreScission™ Protease G allows simultaneous protease immobilization and cleavage of GST fusion proteins.

  • Description: A genetically engineered fusion protein consisting of human rhinovirus 3C protease and GST; Specifically cleaves between the Gln and Gly residues of the recognition sequence of LeuGluValLeuPheGln/GlyPro.
  • Application: Facilitates removal of the protease by allowing simultaneous protease immobilization and cleavage of GST fusion proteins produced from the pGEX-6P vectors pGEX-6P-1, pGEX-6P-2, and pGEX-6P-3
  • Storage Buffer Components: 50mM Tris-HCl (pH 8.0), 150mM NaCl, 10mM EDTA, 1mM DTT, and 20% glycerol.
  • Unit Definition: One unit will cleave >90% of 100μg of a test GST-fusion protein in 50mM Tris-HCl, 150mM NaCl, 1mM EDTA, 1mM DTT, pH 7.0 at 5°C for 16 hrs.

Specifications

Buffer Elution Buffer: 50mM Tris HCl, 10mM reduced Glutathione pH 8; Cleavage Buffer: 50mM Tris HCl pH 8, 150mM NaCl, 10mM EDTA, 1mM DTT and 20% Glycerol
Product Type Fusion Protein Protease
Quantity 500U
Content And Storage -20°C/-80°C
For Use With (Application) Facilitates removal of the protease by allowing simultaneous protease immobilization and cleavage of >90% of 100μg of a test GST-fusion protein produced from the pGEX-6P vectors pGEX-6P-1, pGEX-6P-2 and pGEX-6P-3