missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ preQ1-alkyne
Description
preQ1-alkyne is a modified analog of the natural substrate prequeosine1 (preQ1). It contains alkyne handle that enables users to attach additional groups to preQ1 via Cu(I)-catalyzed azide-alkyne cycloaddition (CuAAC) click chemistry. It can be used to generate photoactivatable probes that can control CRISPR gene editing via the RNA-TAG method.
Other RNA-TAG toolbox compounds include preQ1-biotin ( Cat. No. 7804 )
Plasmids for expression of the E.coli TGT enzyme ( #138201 ) and for cloning an RNA of interest into a vector containing the recognition motif at the 3' end ( #138209 ) are available from Addgene.
Specifications
Specifications
| Chemical Name or Material | N-[6-[[(2-Amino-4,7-dihydro-4-oxo-3H-pyrrolo[2,3-d]pyrimidin-5-yl)methyl]amino]hexyl]-3-(2-propyn-1-yloxy)propanamide |
| CAS | 2771096-76-3 |
| Quantity | 10 mg |
| Target | CRISPR Reagents |
| Molecular Formula | C19H28N6O3 |