missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ PrepSEQ™ Residual DNA Sample Preparation Kit

Catalog No. 50591735
Encompass
Change view
Click to view available options
Quantity:
100 preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50591735 100 preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50591735 Supplier Applied Biosystems™ Supplier No. 4413686

The Applied Biosystems PrepSEQ Residual DNA Sample Preparation Kit is optimized for highly efficient DNA recovery from complex mixtures of proteins, buffers, and salts which are typical of samples that need to be quantitated for residual host cell line DNA.

The Applied Biosystems PrepSEQ Residual DNA Sample Preparation Kit is optimized for highly efficient DNA recovery from complex mixtures of proteins, buffers, and salts which are typical of samples that need to be quantitated for residual host cell line DNA. Quantitative and consistent recovery of DNA can be obtained from challenging matrices, including samples that have high protein concentration, low pH, or high salt.

Specifications

Content And Storage Includes buffers, magnetic beads and enzymes for DNA extraction for residual DNA quantitation for 100 reactions
Isolation Technology Magnetic Bead
Scale Mini
Sample Type Biopharmaceutical Production Samples
Final Product Type Genomic DNA
For Use With (Application) Impurity Testing
High-throughput Compatibility Not High-throughput Compatible (Manual), Automated Protocols
No. of Reactions 100 preps
Product Line PrepSEQ™
Quantity 100 preps
Sample Volume 100 μL
Target Genomic DNA
Test Time 2 hr.
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.