missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ PrepSEQ™ Nucleic Acid Sample Preparation Kit

Catalog No. A50485
Change view
Click to view available options
Quantity:
100 preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A50485 100 preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A50485 Supplier Applied Biosystems™ Supplier No. A50485

The Applied Biosystems PrepSEQ Nucleic Acid Sample Preparation Kit is optimized for highly efficient, quantitative, and consistent recovery of DNA and RNA from diverse samples types.

The Applied Biosystems PrepSEQ Nucleic Acid Sample Preparation Kit is optimized for highly efficient, quantitative, and consistent recovery of DNA and RNA from diverse samples types, including cell culture, raw material (e.g., serum), and in-process downstream purification samples such as column purification or bulk drug substance (BDS) samples.

  • Time-saving, cost-effective DNA and RNA extraction for diverse sample types
  • Reproducible recovery of DNA and RNA from complex samples
  • Manual and high-throughput automated workflow on KingFisher Flex instrument, isolating up to 24 samples (in triplicate) in 5 hours

Easy to use

The PrepSEQ Nucleic Acid Sample Preparation Kit provides a high-throughput automated solution in combination with the Thermo Scientific KingFisher Flex instrument for low hands-on extraction of DNA and RNA.

Consistent performance

For accurate quantitation of DNA and RNA, extraction consistency must be unaffected by complex matrices. Our advanced magnetic bead-based technology provides highly consistent extraction performance to support reliable quantitation.

Specifications

Isolation Technology Magnetic Bead
Sample Type Biopharmaceutical Production Samples
For Use With (Equipment) KingFisher Flex
For Use With (Application) Impurity Testing
High-throughput Compatibility Not High-throughput Compatible (Manual), Automated Protocols
No. of Reactions 100 Reactions
Product Line PrepSEQ
Quantity 100 preps
Sample Volume 100 μL
Target Genomic D, R, Viral R
Test Time 2 hr.
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.