missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ PrepSEQ™ Lysis Buffer

Catalog No. A29825
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A29825 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A29825 Supplier Applied Biosystems™ Supplier No. A29825

PrepSEQ Lysis Buffer is a component of the PrepSEQ Express Nucleic Acid Extraction Kit, offered separately here for those kit users who do large-quantity nucleic acid extractions and would benefit from additional lysis buffer.

This PrepSEQ Lysis Buffer is used in the large-scale protocol of the PrepSEQ Express Nucleic Acid Extraction Kit (Cat. No. 4466351). It is not necessary for the standard protocol (sufficient lysis buffer is included in the kit's lysis tubes). As with other products in the Pharma Analytics Sample Preparation product line, PrepSEQ Lysis Buffer:

  • Is completely supported by our global manufacturing and distribution centers
  • Has a Certificate of Analysis available
  • Has a shelf-life of 12 months

Specifications

Content And Storage 1 x 50 mL bottle, store at room temperature.
Format Bottle
Product Line PrepSEQ
Quantity 50 mL
Reagent Type Cell Lysis Buffer
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.