missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ PrepSEQ™ Express Nucleic Acid Extraction Kit

Catalog No. 4466351
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4466351 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4466351 Supplier Applied Biosystems™ Supplier No. 4466351*

The PrepSEQ Express Nucleic Acid Extraction Kit contains pre-filled reagents and plastic tubes and tips to perform hands-free extraction of purified nucleic acid from 52 samples.

The PrepSEQ Express Nucleic Acid Extraction Kit contains pre-filled reagents and plastic tubes and tips to perform hands-free extraction of purified nucleic acid from 52 samples.

Specifications

Content And Storage Contains pre-filled reagent cartridges and plastics to perform extraction of DNA and RNA.

Store at room temperature.
Isolation Technology Magnetic Bead
Sample Type Biopharmaceutical Purification Sample, Bulk Drug Substance Sample, Cell Cultures
Final Product Type Genomic DNA, Total RNA
For Use With (Equipment) 7500 Fast System, AutoMate Express™ Nucleic Acid Extraction System
High-throughput Compatibility Automated Protocols
No. of Samples Sufficient for isolation of 52 samples
PCR Method Real-Time Quantitative PCR (qPCR)
Product Line PrepSEQ
Quantity 1 Kit
Shipping Condition Room Temperature
Target Genomic DNA, Total RNA
Test Type ≤ 1 hr.
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.