missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ PrepSEQ™ 1-2-3 Nucleic Acid Extraction Kit
GREENER_CHOICE

Catalog No. 4452222
Change view
Click to view available options
Quantity:
100 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4452222 100 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4452222 Supplier Applied Biosystems™ Supplier No. 4452222

The PrepSEQ 1-2-3 Nucleic Acid Extraction Kit uses magnetic particle-based separation technology to extract DNA from Mycoplasma cells or Mouse Minute Virus (MMV) isolated from a variety of starting material.

The PrepSEQ 1-2-3 Nucleic Acid Extraction Kit uses magnetic particle-based separation technology to extract DNA from Mycoplasma cells or Mouse Minute Virus (MMV) isolated from a variety of starting material, such as infected cell cultures or Mycoplasma liquid cultures.

Using this kit, lysate samples are prepared prior to extracting the target DNA. For samples that contain Mycoplasma, the lysates contain Mycoplasma DNA from the free organisms in the culture media and from organisms released from the infected host cells.

Specifications

Content And Storage • PrepSEQ Proteinase K (store at -15°C to -25°C)
• RNASE Cocktail (-15°C to -25°C)
• PrepSEQ Extraction Kit including lysis buffer, Proteinase K buffer, binding solution, wash and elution buffers (room temperature)
• PrepSEQ Magnetic Beads (room temperature)

Format Kit
Isolation Technology Magnetic Bead
Sample Type Cell Culture Supernatants
Final Product Type Genomic DNA, Viral DNA, Bacterial DNA
For Use With (Application) Cell Culture Contamination Detection
Product Line PrepSEQ
Quantity 100 Preps
Target Genomic DNA
Target Organism Class Mouse Minute Virus (MMV), Mycoplasma spp., Vesivirus

For Research Use Only. Not for use in diagnostic procedures.