Learn More
MP Biomedicals™ Precast Gel Plus, Tris-Gly 4 to 20%
These precast polyacrylamide gels are suitable for both SDS-PAGE and Native-PAGE electrophoresis followed by gel staining or Western blotting.
Supplier: MP Biomedicals™ 08W00008
Description
Precast Gel Plus gels are an efficient and convenient scientific tool for separating peptides and small proteins with clear band resolution. In addition, they are compatible with Bio-Rad Mini Protean Tanks, thus facilitating the use of our gels without the need to replace your current hardware.
- Ready-to-use gels for SDS-PAGE or Native-PAGE applications
- Resilient to mechanical stretching during handling
- Removable comb prevents well-wall breakage
- Outlined and numbered wells facilitate gel loading and tracking
- Easy to store and stable at room temperature for 6 months
- Do not freeze
Chemical Identifiers
| 4°C for 12 Months | |
| Clear | |
| 4 to 20% | |
| 1 mm | |
| Peptides, Proteins | |
| Denaturing, Native | |
| 10 Cassettes | |
| Bio-Rad Mini Protean Tanks |
| MP Bio™ Ultra-Fast Running Buffer for SDS-PAGE | |
| 4 to 20% | |
| Mini | |
| Tris-Glycine | |
| 10 to 250 kDa | |
| Gel Staining SDS-PAGE, Native-PAGE, Blue Native, Western Blot, Acrylamide, Tris-Glycine, Mini Gels, Electrophoresis | |
| Precast Gel |
Specifications
| 4°C for 12 Months | |
| Clear | |
| 4 to 20% | |
| 1 mm | |
| ≤30 μL | |
| 10 to 250 kDa | |
| 15-well | |
| 10 Cassettes | |
| Bio-Rad Mini Protean Tanks |
| MP Bio™ Ultra-Fast Running Buffer for SDS-PAGE | |
| 4 to 20% | |
| Mini | |
| Tris-Glycine | |
| Peptides, Proteins | |
| Denaturing, Native | |
| Gel Staining SDS-PAGE, Native-PAGE, Blue Native, Western Blot, Acrylamide, Tris-Glycine, Mini Gels, Electrophoresis | |
| Precast Gel |
Unless specified otherwise, MP Biomedical™'s products are for research or further manufacturing use only, not for direct human use.