Learn More
MP Biomedicals™ Precast Gel Plus, Tris-Gly 10%
These precast polyacrylamide gels are suitable for both SDS-PAGE and Native-PAGE electrophoresis followed by gel staining or Western blotting.
$204.44 - $204.44
Specifications
| Content And Storage | 4°C for 12 Months |
|---|---|
| Buffer | MP Bio™ Ultra-Fast Running Buffer for SDS-PAGE |
| Color | Clear |
| Concentration | 10% |
| For Use With (Equipment) | Bio-Rad Mini Protean Tanks |
| Catalog Number | Mfr. No. | Wells | Sample Loading Volume | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Wells | Sample Loading Volume | Price | Quantity | ||||
|
ICN08W00001
|
MP Biomedicals™
08W00001 |
10-well | ≤50 μL |
Each for $204.44
|
|
||||
|
ICN08W00002
|
MP Biomedicals™
08W00002 |
15-well | ≤30 μL |
Pack of 10 Each for $204.44
|
|
||||
Description
Precast Gel Plus gels are an efficient and convenient scientific tool for separating peptides and small proteins with clear band resolution. In addition, they are compatible with Bio-Rad Mini Protean Tanks, thus facilitating the use of our gels without the need to replace your current hardware.
- Ready-to-use gels for SDS-PAGE or Native-PAGE applications
- Resilient to mechanical stretching during handling
- Removable comb prevents well-wall breakage
- Outlined and numbered wells facilitate gel loading and tracking
- Easy to store and stable at room temperature for 6 months
- Do not freeze
Specifications
| 4°C for 12 Months | |
| Clear | |
| Bio-Rad Mini Protean Tanks | |
| Mini | |
| Tris-Glycine | |
| Peptides, Proteins | |
| Denaturing, Native | |
| 10 Cassettes |
| MP Bio™ Ultra-Fast Running Buffer for SDS-PAGE | |
| 10% | |
| 10% | |
| 1 mm | |
| Precast Gel | |
| 20 to 180 kDa | |
| Gel Staining SDS-PAGE, Native-PAGE, Blue Native, Western Blot, Acrylamide, Tris-Glycine, Mini Gels, Electrophoresis |
Unless specified otherwise, MP Biomedical™'s products are for research or further manufacturing use only, not for direct human use.