missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Precast Gel Plus, Bis-Tris, 4 to 20% Acrylamide, 10 Wells

Catalog No. MP08W00022
Change view
Click to view available options
Wells:
10-well
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Wells
MP08W00022 10-well
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP08W00022 Supplier MP Biomedicals™ Supplier No. 08W00022

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Precast gel optimal for Bis-Tris PAGE Electrophoresis, resilient to mechanical stretching during handling, and featuring outlined and numbered wells for easy loading tracking.

Our Precast Gel Plus is an efficient and convenient scientific tool for use in biological and biochemical fields of study. Perfect for the separation of peptides and small proteins, with great resolution and without slowing down the electrophoresis running times. It allows for a wide range of proteomic analysis. Albeit protein expression, protein cleavage, receptor activation or protein phosphorylation, they can be tracked using established protocols such as Bis-Tris PAGE followed by gel staining or western blotting. In addition, they are also compatible with Bio-Rad Mini Protean Tanks, thus facilitating the use of our gels without the need to replace your hardware.

Used for protein electrophoresis via Bis-Tris PAGE protocols followed by immunoblotting or gel staining. These gels are more robust and can withstand more mechanical stress than the average polyacrylamide gel in the market.

Specifications

Content And Storage 4°C to 8°C
Gel Percentage 4 to 20%
Wells 10-well
Quantity 10 Cassettes

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.