missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Pramipexole dihydrochloride

Catalog No. 417410
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
417410 10 mg
417450 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 417410 Supplier Tocris Bioscience™ Supplier No. 4174/10

Selective D3 agonist

Dopamine receptor agonist exhibiting selectivity for the D3 receptor (Ki values are 0.5, 3.3, 3.9 and 3.9 nM for D3, D2S, D2L and D4 receptors respectively). Exhibits negligible affinity for D1 and D5 receptors. Displays activity in the treatment of Parkinson's disease.

Chemical Identifiers

CAS 104632-25-9
Molecular Formula C10H19Cl2N3S

Specifications

Quantity 10 mg
Formula Weight 284.25
Purity >99%
Molecular Weight (g/mol) 284.243
Chemical Name or Material Pramipexole dihydrochloride
Recommended Storage Desiccate at Room Temperature