missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ PP 242

Catalog No. 42575
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42575 5 mg
425725 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 42575 Supplier Tocris Bioscience™ Supplier No. 4257/5

Dual mTORC1/mTORC2 inhibitor

PP 242 is an ATP-competitive mTORC1/mTORC2 inhibitor (IC50 = 8 nM). Displays selectivity for mTOR over other PI 3K family kinases (IC50 values are 0.102, 0.408, 1.27, 1.96 and 2.2 μM for p110γ, DNA-PK, p110δ, p110α and p110β respectively) and 215 further kinases. Displays modest inhibition of PKCα, JAK2, PKCβI, PKCβII and RET (IC50 values are 0.049, 0.110, 0.185, 0.198 and 0.224 μM respectively). Inhibits both S6K and 4EBP1 phosphorylation; activity causes a decrease in cap-dependent protein translation. Also triggers downregulation of cFLIPS and augments TRAIL-induced apoptosis of cancer cells. PP 242 also inhibits oncogenic K-Ras and PI 3-K induced lipogenesis in breast cancer cells.

Specifications

Chemical Name or Material 2-[4-Amino-1-(1-methylethyl)-1H-pyrazolo[3,4-d]pyrimidin-3-yl]-1H-indol-5-ol
CAS 1092351-67-1
Assay Percent Range >98%
Quantity 5 mg
Molecular Formula C16H16N6O
Formula Weight Observed MW: 308.34