missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel Art™ Poxygrid Test Tube Rack
Chemical and corrosion-resistant steel wire racks are coated with heat-cured epoxy. Orange racks can be used for biohazard warning, yellow for radioactive warning
$290.77 - $355.10
Specifications
| Autoclavable | Autoclavable |
|---|---|
| Color | Blue |
| Holds | Test Tubes |
| Material | Steel Wire |
| Shape | Rectangular |
| Catalog Number | Mfr. No. | Positions | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Positions | Price | Quantity & Availability | |||||
|
03411842
|
Bel Art™
F187820001 |
24 |
Each for $290.77
Case of 24 Each for $6,978.48
|
|
|||||
|
03411843
|
Bel Art™
F187840001 |
48 |
Each for $355.10
Case of 24 Each for $8,522.40
|
|
|||||
Description
- Epoxy coatings are unaffected by organic solvents, salts, weak acids and weak alkalis
- Will withstand a temperature range of -223°C (-369°F) to 121°C (250°F); autoclavable
- All racks have square holes, three decks, a slotted grid bottom and reinforced edges
Specifications
| Autoclavable | |
| Test Tubes | |
| Rectangular | |
| Non-sterile | |
| Test Tube Rack | |
| 10.2 cm |
| Blue | |
| Steel Wire | |
| Yes, When Empty | |
| 1 Ea. | |
| 3 Tiers |