missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ PowerUp™ SYBR™ Green & Endura™ 384-well Plates Bundle

Catalog No. A46940
Encompass
Change view
Click to view available options
Sample Type:
DNA
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
A46940 DNA
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A46940 Supplier Applied Biosystems™ Supplier No. A46940
Get Up to $3,000 of qPCR Products with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Bundle and save up to 50% off PowerUp SYBR Green Master Mix and Plates

  • Reliable performance
  • Highly reproducible
  • Compatible with Applied Biosystems real-time PCR instruments

Specifications

Content And Storage PowerUp™ SYBR™ Green (5 x 5 mL) and 20 Endura™ 384-well Plates. Store at 2–8°C
Detection Method SYBR
For Use With (Application) Real Time PCR (qPCR)
Sample Type DNA