Learn More
Treston™ Power Rail with Upright Mounting Bracket
Mounting positions include horizontal and vertical on uprights, rear case mounting, front rail mounting, and work surface mounting. Power rails are available with surge protection.
$540.32 - $721.72
| Catalog Number | Mfr. No. | Includes | Description | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Includes | Description | Price | Quantity | ||||
|
1491118807
|
Treston™
1491118807 |
6 Outlets | M30 Power Rail |
Each for $540.32
|
|
||||
|
1491118808
|
Treston™
1491118808 |
6 Outlets, Surge | M30 Power Rail |
Each for $617.19
|
|
||||
|
1491118809
|
Treston™
1491118809 |
8 Outlets | M36 Power Rail |
Each for $595.66
|
|
||||
|
1491118810
|
Treston™
1491118810 |
8 Outlets, Surge | M36 Power Rail |
Each for $672.53
|
|
||||
|
1491118811
|
Treston™
1491118811 |
10 Outlets | M48 Power Rail |
Each for $644.85
|
|
||||
|
1491118812
|
Treston™
1491118812 |
10 Outlets, Surge | M48 Power Rail |
Each for $721.72
|
|
||||
Description
- 20 amp, 125 volt, NEMA 5-20P plug
- Black aluminum housing with on/off switch
- Upright mounting brackets included for horizontal or vertical mounting positions