missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Treston™ Power Rail with Upright Mounting Bracket
Mounting positions include horizontal and vertical on uprights, rear case mounting, front rail mounting, and work surface mounting. Power rails are available with surge protection.
Supplier: Treston™ 1491118808
Description
- 20 amp, 125 volt, NEMA 5-20P plug
- Black aluminum housing with on/off switch
- Upright mounting brackets included for horizontal or vertical mounting positions
Specifications
| 24.5 in. | |
| Power Rail | |
| 51 mm | |
| 1 Ea. | |
| 24.5 in. | |
| 622 mm | |
| 622 mm |
| M30 Power Rail | |
| 2 in. | |
| 2 in. | |
| 6 Outlets, Surge | |
| 51 mm | |
| Treston Workstations | |
| Steel |