missing translation for 'onlineSavingsMsg'
Learn More

Potassium (trifluoromethyl)trifluoroborate, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 464730050

 View more versions of this product

Catalog No. AC464730050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Potassium (trifluoromethyl)trifluoroborate
CBF6K
Solubility aq. soln.: soluble.
[K+].F[B-](F)(F)C(F)(F)F
175.91
42298-15-7
5 g
UOGBGCVSVMQVBR-UHFFFAOYSA-N
potassium trifluoro(trifluoromethyl)boranuide
175.91

Chemical Identifiers

42298-15-7
175.91
potassium trifluoro(trifluoromethyl)boranuide
CBF6K
UOGBGCVSVMQVBR-UHFFFAOYSA-N
[K+].F[B-](F)(F)C(F)(F)F
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement Harmful if swallowed.

Harmful in contact with skin.

Causes skin irritation.

Causes serious eye irritation.

Harmful if inhaled.

May cause respiratory irritation.

GHS P Statement IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Call a POISON CENTER or doctor/physician if you feel unwell.

IF ON SKIN: Wash with plenty of soap and water.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

If eye irritation persists: Get medical advice/attention.

Wear protective gloves/protective clothing/eye protection/face protection.

If skin irritation occurs: Get medical advice/attention.

GHS Signal Word: Warning

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only