accessibility menu, dialog, popup

UserName

CAS RN 42298-15-7

IUPAC Name: potassium trifluoro(trifluoromethyl)boranuide
Synonyms: potassium trifluoro(trifluoromethyl)boranuide
Molecular Weight (g/mol): 175.91
Molecular Formula: CBF6K
InChi Key: UOGBGCVSVMQVBR-UHFFFAOYSA-N
SMILES: [K+].F[B-](F)(F)C(F)(F)F
Quantity 
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (3)