Learn More
Potassium (trifluoromethyl)trifluoroborate, Thermo Scientific™
Chemical Identifiers
| CAS | 42298-15-7 |
|---|---|
| Molecular Formula | CBF6K |
| Molecular Weight (g/mol) | 175.91 |
| InChI Key | UOGBGCVSVMQVBR-UHFFFAOYSA-N |
| IUPAC Name | potassium trifluoro(trifluoromethyl)boranuide |
| SMILES | [K+].F[B-](F)(F)C(F)(F)F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AC464730010
|
N/A | 1 g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC464730050
|
N/A | 5 g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 42298-15-7 | |
| 175.91 | |
| potassium trifluoro(trifluoromethyl)boranuide |
| CBF6K | |
| UOGBGCVSVMQVBR-UHFFFAOYSA-N | |
| [K+].F[B-](F)(F)C(F)(F)F |
Specifications
| 42298-15-7 | |
| Solubility aq. soln.: soluble. | |
| [K+].F[B-](F)(F)C(F)(F)F | |
| 175.91 | |
| Potassium (trifluoromethyl)trifluoroborate |
| CBF6K | |
| UOGBGCVSVMQVBR-UHFFFAOYSA-N | |
| potassium trifluoro(trifluoromethyl)boranuide | |
| 175.91 |
Safety and Handling
GHS P Statement IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
GHS Signal Word: Warning
RUO – Research Use Only