Learn More
Potassium (trifluoromethyl)trifluoroborate, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 464730010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| Potassium (trifluoromethyl)trifluoroborate | |
| CBF6K | |
| Solubility aq. soln.: soluble. | |
| [K+].F[B-](F)(F)C(F)(F)F | |
| 175.91 |
| 42298-15-7 | |
| 1 g | |
| UOGBGCVSVMQVBR-UHFFFAOYSA-N | |
| potassium trifluoro(trifluoromethyl)boranuide | |
| 175.91 |
Chemical Identifiers
| 42298-15-7 | |
| 175.91 | |
| potassium trifluoro(trifluoromethyl)boranuide |
| CBF6K | |
| UOGBGCVSVMQVBR-UHFFFAOYSA-N | |
| [K+].F[B-](F)(F)C(F)(F)F |
Safety and Handling
GHS P Statement IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
GHS Signal Word: Warning
RUO â Research Use Only