missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium titanium oxide
CAS: 12030-97-6 | K2O3Ti | 174.06 g/mol
$106.97 - $346.00
Chemical Identifiers
| CAS | 12030-97-6 |
|---|---|
| Molecular Formula | K2O3Ti |
| Molecular Weight (g/mol) | 174.06 |
| MDL Number | MFCD00058803 |
| InChI Key | KEWYKJOOCFOYTD-UHFFFAOYSA-N |
| Synonym | kaliumtitanat, titanate,dipotassium, potassiumtitanatewhisker, 2k.o3ti, titanate tio32-,dipotassium, dipotassium;dioxido oxo titanium |
| PubChem CID | 45052161 |
| SMILES | [O--].[O--].[O--].[K+].[K+].[Ti+4] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2026322
|
Thermo Scientific Chemicals
B2026322 |
100 g |
Each for $106.97
|
|
|||||
|
AAB2026336
|
Thermo Scientific Chemicals
B2026336 |
500 g |
Each for $346.00
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 12030-97-6 | |
| 174.06 | |
| KEWYKJOOCFOYTD-UHFFFAOYSA-N | |
| 45052161 |
| K2O3Ti | |
| MFCD00058803 | |
| kaliumtitanat, titanate,dipotassium, potassiumtitanatewhisker, 2k.o3ti, titanate tio32-,dipotassium, dipotassium;dioxido oxo titanium | |
| [O--].[O--].[O--].[K+].[K+].[Ti+4] |
Specifications
| 1615°C | |
| 100 g | |
| MFCD00058803 | |
| KEWYKJOOCFOYTD-UHFFFAOYSA-N | |
| dipotassium;dioxido(oxo)titanium | |
| 45052161 | |
| Potassium titanium oxide |
| 12030-97-6 | |
| K2O3Ti | |
| kaliumtitanat, titanate,dipotassium, potassiumtitanatewhisker, 2k.o3ti, titanate tio32-,dipotassium, dipotassium;dioxido oxo titanium | |
| [O--].[O--].[O--].[K+].[K+].[Ti+4] | |
| 174.06 | |
| Odorless |
Safety and Handling
EINECSNumber : 234-748-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only