accessibility menu, dialog, popup

UserName

CAS RN 12030-97-6

OOOKKTi

CAS RN 12030-97-6

IUPAC Name: titanium(4+) dipotassium trioxidandiide
Synonyms: potassium titanium oxide (K2ti6O13)
Molecular Weight (g/mol): 174.06
Molecular Formula: K2O3Ti
InChi Key: KEWYKJOOCFOYTD-UHFFFAOYSA-N
SMILES: [O--].[O--].[O--].[K+].[K+].[Ti+4]
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results