missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium titanium oxide
CAS: 12030-97-6 | K2O3Ti | 174.06 g/mol
Supplier: Thermo Scientific Chemicals B2026336
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Potassium titanium oxide | |
| 12030-97-6 | |
| K2O3Ti | |
| kaliumtitanat, titanate,dipotassium, potassiumtitanatewhisker, 2k.o3ti, titanate tio32-,dipotassium, dipotassium;dioxido oxo titanium | |
| [O--].[O--].[O--].[K+].[K+].[Ti+4] | |
| 174.06 | |
| Odorless |
| 1615°C | |
| 500 g | |
| MFCD00058803 | |
| KEWYKJOOCFOYTD-UHFFFAOYSA-N | |
| dipotassium;dioxido(oxo)titanium | |
| 45052161 |
Chemical Identifiers
| 12030-97-6 | |
| 174.06 | |
| KEWYKJOOCFOYTD-UHFFFAOYSA-N | |
| 45052161 |
| K2O3Ti | |
| MFCD00058803 | |
| kaliumtitanat, titanate,dipotassium, potassiumtitanatewhisker, 2k.o3ti, titanate tio32-,dipotassium, dipotassium;dioxido oxo titanium | |
| [O--].[O--].[O--].[K+].[K+].[Ti+4] |
Safety and Handling
EINECSNumber : 234-748-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only