missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium thiocyanate, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99%, Honeywell Fluka™
Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur.,≥99%
Supplier: Honeywell Chemicals 3127225KG
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
| Potassium Thiocyanate | |
| 333-20-0 | |
| CKNS | |
| MFCD00011413 | |
| 3594799 | |
| ZNNZYHKDIALBAK-UHFFFAOYSA-M | |
| potassium;thiocyanate | |
| 516872 | |
| 97.18g/mol | |
| ACS/puriss. p.a. |
| Reag. Ph. Eur. reag. ISO | |
| 2.5 kg | |
| KSCN | |
| NONH for all modes of transport | |
| potassium thiocyanate, potassium rhodanate, potassium rhodanide, rodanca, potassium sulfocyanate, rhodanide, rhocya, potassium thiocyanide, kscn, thiocyanic acid, potassium salt | |
| C(#N)[S-].[K+] | |
| 97.176 | |
| CHEBI:30951 | |
| ≥99% |
Chemical Identifiers
| 333-20-0 | |
| 97.176 | |
| ZNNZYHKDIALBAK-UHFFFAOYSA-M | |
| 516872 | |
| potassium;thiocyanate |
| CKNS | |
| MFCD00011413 | |
| potassium thiocyanate, potassium rhodanate, potassium rhodanide, rodanca, potassium sulfocyanate, rhodanide, rhocya, potassium thiocyanide, kscn, thiocyanic acid, potassium salt | |
| CHEBI:30951 | |
| C(#N)[S-].[K+] |
Safety and Handling
P273-P280
H302 + H312 + H332-H412
EINECSNumber : 206-370-1