missing translation for 'onlineSavingsMsg'
Learn More

Potassium thiocyanate, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99%, Honeywell Fluka™

Catalog No. p-7072019
Change view
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
6 x 250 g
6 x 1 kg
6 x 2.5 kg
Catalog No. Quantity
6002344 250 g
6002342 1 kg
60045940 6 x 250 g
6002343 2.5 kg
60045938 6 x 1 kg
60045939 6 x 2.5 kg
6 options

Catalog No. 6002344

Supplier: Honeywell Chemicals 31272250G

Only null left

Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur.,≥99%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 333-20-0
Molecular Formula CKNS
Molecular Weight (g/mol) 97.176
MDL Number MFCD00011413
InChI Key ZNNZYHKDIALBAK-UHFFFAOYSA-M
Synonym potassium thiocyanate, potassium rhodanate, potassium rhodanide, rodanca, potassium sulfocyanate, rhodanide, rhocya, potassium thiocyanide, kscn, thiocyanic acid, potassium salt
PubChem CID 516872
ChEBI CHEBI:30951
IUPAC Name potassium;thiocyanate
SMILES C(#N)[S-].[K+]

Specifications

Quantity 250 g
Linear Formula KSCN
UN Number NONH for all modes of transport
Beilstein 3594799
Formula Weight 97.18g/mol
Percent Purity ≥99%
Grade ACS/puriss. p.a.
Chemical Name or Material Potassium Thiocyanate, Reag. Ph. Eur. reag. ISO
Hazard Statement P273-P280
Precautionary Statement H302 + H312 + H332-H412
EINECSNumber 206-370-1