missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium thiocyanate concentrate, For 1L standard solution, 0.1 M KSCN, Honeywell Fluka™
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| CAS Min % | 80.0000%, 10.0000% |
| CAS Max % | 90.0000%, 20.0000% |
| Physical Form | Concentrate |
| Quantity | 6 x 1 Ea. |
| Linear Formula | KSCN |
| UN Number | NONH for all modes of transport |
| Beilstein | 3594799 |
| Formula Weight | 97.18 |
| Packaging | Ampoule |
| Density | 1 g/mL at 25 °C(lit.) |
| Show More |
Safety and Handling
| ShelfLife | 1440 days from date of manufacture |
| Recommended Storage | Room Temp |