missing translation for 'onlineSavingsMsg'
Learn More

Potassium Thiocyanate, Certified, 10.0% (w/v) ±0.4%, LabChem™

Catalog No. LC221401
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC221401 500 mL
LC221402 1 L
LC221404 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. LC221401 Supplier LabChem Supplier No. LC221401

Chemical Identifiers

CAS 333-20-0 , 7732-18-5
Molecular Formula CKNS
Molecular Weight (g/mol) 97.176
InChI Key ZNNZYHKDIALBAK-UHFFFAOYSA-M
Synonym potassium thiocyanate, potassium rhodanate, potassium rhodanide, rodanca, potassium sulfocyanate, rhodanide, rhocya, potassium thiocyanide, kscn, thiocyanic acid, potassium salt
PubChem CID 516872
ChEBI CHEBI:30951
IUPAC Name potassium;thiocyanate
SMILES C(#N)[S-].[K+]

Specifications

Color Colorless
CAS Max % 90, 10
Physical Form Liquid
Quantity 500 mL
Linear Formula KSCN
Solubility Information Soluble in water
Formula Weight 97.18
Percent Purity 10.0% ±0.4% (w/v)
Grade Certified
Identification Passes Test
Packaging Poly Bottle
Decomposition Information Hydrogen cyanide; Nitrogen oxides; Sulfur compounds; Carbon monoxide; Carbon dioxide
Chemical Name or Material Potassium Thiocyanate
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Recommended Storage Room Temperature