missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium tert-Butoxide 97.0+%, TCI America™
Supplier: TCI America P1008500G
Specifications
| Potassium tert-Butoxide | |
| 240°C | |
| C4H9KO | |
| 500 g | |
| potassium tert-butoxide, potassium tert-butanolate, potassium t-butoxide, potassium 2-methylpropan-2-olate, potassium tert-butylate, kotbu, 2-methyl-2-propanol, potassium salt, tert-butoxypotassium, potassium-t-butoxide, t-buok | |
| CC(C)(C)[O-].[K+] | |
| 112.213 | |
| 112.21 | |
| Crystalline Powder |
| 865-47-4 | |
| White-Yellow | |
| MFCD00012162 | |
| 3206 | |
| LPNYRYFBWFDTMA-UHFFFAOYSA-N | |
| potassium;2-methylpropan-2-olate | |
| 23665647 | |
| ≥97.0% (T) |
Chemical Identifiers
| 865-47-4 | |
| 112.213 | |
| LPNYRYFBWFDTMA-UHFFFAOYSA-N | |
| 23665647 | |
| CC(C)(C)[O-].[K+] |
| C4H9KO | |
| MFCD00012162 | |
| potassium tert-butoxide, potassium tert-butanolate, potassium t-butoxide, potassium 2-methylpropan-2-olate, potassium tert-butylate, kotbu, 2-methyl-2-propanol, potassium salt, tert-butoxypotassium, potassium-t-butoxide, t-buok | |
| potassium;2-methylpropan-2-olate |
Safety and Handling
EINECSNumber : (2)-3100
TSCA : Yes