missing translation for 'onlineSavingsMsg'
Learn More

Potassium tert-Butoxide 97.0+%, TCI America™

Catalog No. p-7134897
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Catalog No. Quantity
P100825G 25 g
P1008100G 100 g
P1008500G 500 g
3 options

Catalog No. P100825G

Supplier: TCI America P100825G

Only null left

Chemical Identifiers

CAS 865-47-4
Molecular Formula C4H9KO
Molecular Weight (g/mol) 112.213
MDL Number MFCD00012162
InChI Key LPNYRYFBWFDTMA-UHFFFAOYSA-N
Synonym potassium tert-butoxide, potassium tert-butanolate, potassium t-butoxide, potassium 2-methylpropan-2-olate, potassium tert-butylate, kotbu, 2-methyl-2-propanol, potassium salt, tert-butoxypotassium, potassium-t-butoxide, t-buok
PubChem CID 23665647
IUPAC Name potassium;2-methylpropan-2-olate
SMILES CC(C)(C)[O-].[K+]

Specifications

Melting Point 240°C
Color White-Yellow
Quantity 25 g
UN Number 3206
Formula Weight 112.21
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Potassium tert-Butoxide
EINECSNumber (2)-3100
TSCA Yes