CAS RN 865-47-4
Potassium tert-butoxide, 1M solution in tert-butanol, AcroSeal™
CAS: 865-47-4 Molecular Formula: C4H9KO Molecular Weight (g/mol): 112.21 MDL Number: MFCD00012162 InChI Key: LPNYRYFBWFDTMA-UHFFFAOYSA-N Synonym: potassium tert-butoxide,potassium tert-butanolate,potassium t-butoxide,potassium 2-methylpropan-2-olate,potassium tert-butylate,kotbu,2-methyl-2-propanol, potassium salt,tert-butoxypotassium,potassium-t-butoxide,t-buok PubChem CID: 23665647 IUPAC Name: potassium;2-methylpropan-2-olate SMILES: CC(C)(C)[O-].[K+]
Potassium tert-butoxide, pure, 20 wt.% solution in THF
CAS: 865-47-4 | C4H9KO | 112.21 g/mol
Potassium tert-butoxide, 2M (25% w/w) solution in 2-MeTHF, AcroSeal™
CAS: 865-47-4 | C4H9KO | 112.21 g/mol
Potassium tert-butoxide, 98+%, pure
CAS: 865-47-4 Molecular Formula: C4H9KO Molecular Weight (g/mol): 112.21 InChI Key: LPNYRYFBWFDTMA-UHFFFAOYSA-N Synonym: potassium tert-butoxide,potassium tert-butanolate,potassium t-butoxide,potassium 2-methylpropan-2-olate,potassium tert-butylate,kotbu,2-methyl-2-propanol, potassium salt,tert-butoxypotassium,potassium-t-butoxide,t-buok PubChem CID: 23665647 IUPAC Name: potassium;2-methylpropan-2-olate SMILES: CC(C)(C)[O-].[K+]
Potassium tert-butoxide, pure, 1.6-1.7M (20 wt.%) solution in THF, AcroSeal™
CAS: 865-47-4 | C4H9KO | 112.21 g/mol
Potassium tert-butoxide, pure, 1M solution in THF, AcroSeal™
CAS: 865-47-4 | C4H9KO | 112.21 g/mol
Potassium tert-butoxide, 97%
CAS: 865-47-4 Molecular Formula: C4H9KO Molecular Weight (g/mol): 112.213 MDL Number: MFCD00012162 InChI Key: LPNYRYFBWFDTMA-UHFFFAOYSA-N Synonym: potassium tert-butoxide,potassium tert-butanolate,potassium t-butoxide,potassium 2-methylpropan-2-olate,potassium tert-butylate,kotbu,2-methyl-2-propanol, potassium salt,tert-butoxypotassium,potassium-t-butoxide,t-buok PubChem CID: 23665647 IUPAC Name: potassium;2-methylpropan-2-olate SMILES: CC(C)(C)[O-].[K+]
Potassium tert-Butoxide 97.0+%, TCI America™
CAS: 865-47-4 Molecular Formula: C4H9KO Molecular Weight (g/mol): 112.213 MDL Number: MFCD00012162 InChI Key: LPNYRYFBWFDTMA-UHFFFAOYSA-N Synonym: potassium tert-butoxide,potassium tert-butanolate,potassium t-butoxide,potassium 2-methylpropan-2-olate,potassium tert-butylate,kotbu,2-methyl-2-propanol, potassium salt,tert-butoxypotassium,potassium-t-butoxide,t-buok PubChem CID: 23665647 IUPAC Name: potassium;2-methylpropan-2-olate SMILES: CC(C)(C)[O-].[K+]
Potassium tert-Butoxide (12% in Tetrahydrofuran, ca. 1mol/L), TCI America™
CAS: 865-47-4 Molecular Formula: C4H9KO Molecular Weight (g/mol): 112.213 InChI Key: LPNYRYFBWFDTMA-UHFFFAOYSA-N Synonym: potassium tert-butoxide,potassium tert-butanolate,potassium t-butoxide,potassium 2-methylpropan-2-olate,potassium tert-butylate,kotbu,2-methyl-2-propanol, potassium salt,tert-butoxypotassium,potassium-t-butoxide,t-buok PubChem CID: 23665647 IUPAC Name: potassium;2-methylpropan-2-olate SMILES: CC(C)(C)[O-].[K+]
Sigma Aldrich Potassium tert-butoxide solution
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.