missing translation for 'onlineSavingsMsg'
Learn More

Potassium Sulfate TS, 1% (w/v), Ricca Chemical

$147.33 - $147.33

Chemical Identifiers

CAS 7778-80-5
Molecular Formula K2O4S
Molecular Weight (g/mol) 174.25
MDL Number MFCD00011388
InChI Key OTYBMLCTZGSZBG-UHFFFAOYSA-L
Synonym potassium sulfate, dipotassium sulfate, potassium sulphate, sulfuric acid dipotassium salt, sal polychrestum, sulfuric acid, potassium salt, arcanum duplicatum, kalium sulphuricum, caswell no. 702, sulfuric acid, dipotassium salt
PubChem CID 24507
ChEBI CHEBI:32036
IUPAC Name dipotassium sulfate
SMILES [K+].[K+].[O-]S([O-])(=O)=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
6540-4
View Documents
Ricca Chemical Company
RI65404
120 mL Natural Poly Bottle
Each for $147.33
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7778-80-5
174.25
OTYBMLCTZGSZBG-UHFFFAOYSA-L
24507
dipotassium sulfate
K2O4S
MFCD00011388
potassium sulfate, dipotassium sulfate, potassium sulphate, sulfuric acid dipotassium salt, sal polychrestum, sulfuric acid, potassium salt, arcanum duplicatum, kalium sulphuricum, caswell no. 702, sulfuric acid, dipotassium salt
CHEBI:32036
[K+].[K+].[O-]S([O-])(=O)=O

Specifications

Colorless
97.0,0.97
K2O4S
potassium sulfate, dipotassium sulfate, potassium sulphate, sulfuric acid dipotassium salt, sal polychrestum, sulfuric acid, potassium salt, arcanum duplicatum, kalium sulphuricum, caswell no. 702, sulfuric acid, dipotassium salt
[K+].[K+].[O-]S([O-])(=O)=O
174.25
CHEBI:32036
Laboratory
Aqueous Solution
120 mL
1% (w/v)
7778-80-5,7732-18-5
100.0,1.01
MFCD00011388
OTYBMLCTZGSZBG-UHFFFAOYSA-L
dipotassium sulfate
24507
1%
Natural Poly Bottle
∼7
Liquid
Potassium Sulfate, Test Solution
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'