missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Sulfate, Crystal, ACS, 99%, Spectrum™ Chemical
$249.41 - $9293.34
Chemical Identifiers
| CAS | 7778-80-5 |
|---|---|
| Molecular Formula | K2O4S |
| Molecular Weight (g/mol) | 174.25 |
| MDL Number | MFCD00011388 |
| InChI Key | OTYBMLCTZGSZBG-UHFFFAOYSA-L |
| IUPAC Name | dipotassium sulfate |
| SMILES | [K+].[K+].[O-]S([O-])(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18612906
|
Spectrum Chemical Mfg Cor
P1405125GM |
125 g | Poly Bottle |
Each for $249.41
|
|
||||
|
18612907
|
Spectrum Chemical Mfg Cor
P1405500GM |
500 g | Poly Bottle |
Each for $461.13
|
|
||||
|
18612908
|
Spectrum Chemical Mfg Cor
P140525KG |
2.5 kg | Poly Bottle |
Each for $1,536.00
|
|
||||
|
18612909
|
Spectrum Chemical Mfg Cor
P140512KG |
12 kg | Poly Pail |
Each for $4,116.47
|
|
||||
|
18612910
|
Spectrum Chemical Mfg Cor
P140550KGBL |
50 kg | Fiber Drum |
Each for $9,293.34
|
|
||||
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.