missing translation for 'onlineSavingsMsg'
Learn More

Potassium Sulfate, Crystal, ACS, 99%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor P140512KG

 View more versions of this product

Catalog No. 18612909


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

7778-80-5
5.5 to 8.5
K2O4S
OTYBMLCTZGSZBG-UHFFFAOYSA-L
dipotassium sulfate
99%
Poly Pail
1
12 kg
MFCD00011388
[K+].[K+].[O-]S([O-])(=O)=O
174.25
ACS

Chemical Identifiers

7778-80-5
174.25
OTYBMLCTZGSZBG-UHFFFAOYSA-L
[K+].[K+].[O-]S([O-])(=O)=O
K2O4S
MFCD00011388
dipotassium sulfate
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.