missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Sorbate, Granular, NF, 98-101%, Spectrum™ Chemical
PO301, 590-00-1, C6H7KO2
$299.67 - $15475.70
Chemical Identifiers
| CAS | 590-00-1 |
|---|---|
| Molecular Formula | C6H7KO2 |
| Molecular Weight (g/mol) | 150.22 |
| MDL Number | MFCD00016546 |
| InChI Key | CHHHXKFHOYLYRE-STWYSWDKSA-M |
| IUPAC Name | potassium (2E,4E)-hexa-2,4-dienoate |
| SMILES | [K+].C\C=C\C=C\C([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18605711
|
Spectrum Chemical Mfg Cor
PO301125GM |
125 g | Amber Glass Bottle |
Each for $299.67
|
|
||||
|
18605712
|
Spectrum Chemical Mfg Cor
PO301500GM |
500 g | Amber Glass Bottle |
Each for $700.40
|
|
||||
|
18605713
|
Spectrum Chemical Mfg Cor
PO30125KG |
2.5 kg | Poly Pail |
Each for $1,895.23
|
|
||||
|
18605714
|
Spectrum Chemical Mfg Cor
PO30112KG |
12 kg | Fiber Drum |
Each for $4,729.27
|
|
||||
|
18605715
|
Spectrum Chemical Mfg Cor
PO30150KGBL |
50 kg | Fiber Drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical Potassium Sorbate, Granular, NF is used as an antimicrobial preservative. All Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
| 590-00-1 | |
| 150.22 | |
| CHHHXKFHOYLYRE-STWYSWDKSA-M | |
| [K+].C\C=C\C=C\C([O-])=O |
| C6H7KO2 | |
| MFCD00016546 | |
| potassium (2E,4E)-hexa-2,4-dienoate |
Specifications
| 590-00-1 | |
| 125 g | |
| MFCD00016546 | |
| [K+].C\C=C\C=C\C([O-])=O | |
| 150.22 | |
| NF | |
| Amber Glass Bottle |
| 1 | |
| C6H7KO2 | |
| CHHHXKFHOYLYRE-STWYSWDKSA-M | |
| potassium (2E,4E)-hexa-2,4-dienoate | |
| 98 to 101% | |
| 0.01 |