missing translation for 'onlineSavingsMsg'
Learn More

Potassium Sorbate, Granular, NF, 98-101%, Spectrum™ Chemical

Catalog No. 18605715
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18605715 50 kg Fiber Drum
18605711 125 g Amber Glass Bottle
18605712 500 g Amber Glass Bottle
18605713 2.5 kg Poly Pail
18605714 12 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18605715 Supplier Spectrum Chemical Mfg Cor Supplier No. PO30150KGBL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

PO301, 590-00-1, C6H7KO2

Spectrum™ Chemical Potassium Sorbate, Granular, NF is used as an antimicrobial preservative. All Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 590-00-1
Molecular Formula C6H7KO2
Molecular Weight (g/mol) 150.22
MDL Number MFCD00016546
InChI Key CHHHXKFHOYLYRE-STWYSWDKSA-M
IUPAC Name potassium (2E,4E)-hexa-2,4-dienoate
SMILES [K+].C\C=C\C=C\C([O-])=O

Specifications

CAS Max % 1
Quantity 50 kg
Percent Purity 98 to 101%
Grade NF
Loss on Drying 0.01
Packaging Fiber Drum