missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Sorbate, Granular, NF, 98-101%, Spectrum™ Chemical
PO301, 590-00-1, C6H7KO2
Supplier: Spectrum Chemical Mfg Cor PO30112KG
Description
Spectrum™ Chemical Potassium Sorbate, Granular, NF is used as an antimicrobial preservative. All Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
| 590-00-1 | |
| 12 kg | |
| MFCD00016546 | |
| [K+].C\C=C\C=C\C([O-])=O | |
| 150.22 | |
| NF | |
| Fiber Drum |
| 1 | |
| C6H7KO2 | |
| CHHHXKFHOYLYRE-STWYSWDKSA-M | |
| potassium (2E,4E)-hexa-2,4-dienoate | |
| 98 to 101% | |
| 0.01 |
Chemical Identifiers
| 590-00-1 | |
| 150.22 | |
| CHHHXKFHOYLYRE-STWYSWDKSA-M | |
| [K+].C\C=C\C=C\C([O-])=O |
| C6H7KO2 | |
| MFCD00016546 | |
| potassium (2E,4E)-hexa-2,4-dienoate |