missing translation for 'onlineSavingsMsg'
Learn More

Potassium Pyroantimonate TS, Solution, Spectrum™ Chemical

Catalog No. 18605693
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18605693 500 mL Poly Bottle
18605694 1 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18605693 Supplier Spectrum Chemical Mfg Cor Supplier No. P198500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

P-198, 12208-13-8

Spectrum™ Chemical Potassium Pyroantimonate TS, Solution is a solution, sometimes designated with 'TS or Test Solution', that is prepared according to the specifications of the USP Pharmacopeial Convention, for the purpose of conducting USP and National Formulary tests and assays.

Chemical Identifiers

CAS 12208-13-8 , 1310-58-3 , 7732-18-5
Molecular Formula H6KO6Sb
Molecular Weight (g/mol) 262.90
InChI Key IDVSLFDKDQVCAP-UHFFFAOYSA-H
IUPAC Name antimony(3+) potassium hexahydroxide
SMILES [OH-].[OH-].[OH-].[OH-].[OH-].[OH-].[K+].[Sb+3]

Specifications

CAS Max % 0.014, 0.978, 0.018
UN Number UN1814
Grade Chemical Solution
Packaging Poly Bottle
Quantity 500 mL
Chemical Name or Material Potassium Pyroantimonate TS